Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
CNPY2 is positive regulator of neurite outgrowth by stabilizing myosin regulatory light chain (MRLC). It prevents MIR-mediated MRLC ubiquitination and its subsequent proteasomal degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: canopy 2 homolog; MIR-interacting saposin-like protein; Protein canopy homolog 2; Putative secreted protein ZSIG9; Transmembrane protein 4
Gene Aliases: 5330432A10Rik; AW229003; CNPY2; D10Bwg1546e; HP10390; MSAP; TMEM4; UNQ1943/PRO4426; ZSIG9
UniProt ID: (Human) Q9Y2B0, (Mouse) Q9QXT0
Entrez Gene ID: (Human) 10330, (Mouse) 56530, (Rat) 685814
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support