Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEKSRMNLPK GPDTLCFDKD EFMKEDFDVD HFVSDCRKRV QLEELRDDLE LYYKLLKTAM VELINKDYAD FVNLSTNLVG MDKALNQLSV PLGQLREEVL SLRSSVSEGI RAVDERMSKQ EDIRKKKMCV LRLIQVIRSV EKIEKILNSQ SSKETSALEA SSPLLTGQIL ERIATEFNQL QFHAVQSKGM PLLDKVRPRI AGITAMLQQS LEGLLLEGLQ TSDVDIIRHC LRTYATIDKT RDAEALVGQV LVKPYIDEVI IEQFVESHPN GLQVMYNKLL EFVPHHCRLL REVTGGAISS EKGNTVPGYD FLVNSVWPQI VQGLEEKLPS LFNPGNPDAF HEKYTISMDF VRRLERQCGS QASVKRLRAH PAYHSFNKKW NLPVYFQIRF REIAGSLEAA LTDVLEDAPA ESPYCLLASH RTWSSLRRCW SDEMFLPLLV HRLWRLTLQI LARYSVFVNE LSLRPISNES PKEIKKPLVT GSKEPSITQG NTEDQGSGPS ETKPVVSISR TQLVYVVADL DKLQEQLPEL LEIIKPKLEM IGFKNFSSIS AALEDSQSSF SACVPSLSSK IIQDLSDSCF GFLKSALEVP RLYRRTNKEV PTTASSYVDS ALKPLFQLQS GHKDKLKQAI IQQWLEGTLS ESTHKYYETV SDVLNSVKKM EESLKRLKQA RKTTPANPVG PSGGMSDDDK IRLQLALDVE YLGEQIQKLG LQASDIKSFS ALAELVAAAK DQATAEQP
The structure and function of the Golgi apparatus is controlled by a number of multi-protein complexes that are involved in glycosylation reactions and vesicular transport. The conserved oligomeric Golgi (COG) complex consists of three subcomplexes, termed LDLC, SEC34 and GTT (Golgi transport complex), all of which contain proteins necessary for proper Golgi operation. COG2 (conserved oligomeric Golgi complex subunit 2), also known as LDLC, is a 730 amino acid component of the COG complex. Localized to the cytoplasmic side of the Golgi apparatus, COG2 is required for proper Golgi morphology and function, specifically playing a role in Golgi ribbon formation and vesicular transport. Abnormal COG2 function may cause cell death, suggesting that COG2 is an important factor in cell viability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brefeldin A-sensitive, peripheral Golgi protein; COG 2; COG complex subunit 2; Component of oligomeric Golgi complex 2; conserved oligomeric Golgi complex protein 2; Conserved oligomeric Golgi complex subunit 2; low density lipoprotein receptor defect C complementing; Low density lipoprotein receptor defect C-complementing protein; Transcript ch4822
Gene Aliases: COG2; LDLC
UniProt ID: (Human) Q14746
Entrez Gene ID: (Human) 22796
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support