Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total Collagen XIV alpha 1.
Type XIV collagen is a fibril-associated collagen with an interrupted triple helix. It interacts with the fibril surface and regulates fibrillogenesis (Ansorge et al., 2009 [PubMed 19136672]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: col14a1; Collagen alpha-1(XIV) chain; collagen, type XIV, alpha 1; procollagen, type XIV, alpha 1; Undulin; undulin (fibronectin-tenascin-related)
Gene Aliases: 5730412L22Rik; AW108078; COL14A1; UND
UniProt ID: (Human) Q05707, (Mouse) Q80X19
Entrez Gene ID: (Human) 7373, (Mouse) 12818
Molecular Function:
extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support