Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PTGDPGKNGD KGHAGLAGAR GAPGPDGNNG AQGPPGPQGV QGGKGEQGPP GPPGFQGLPG PSGPAGEVGK PGERGLHGEF GLPGPAGPRG ERGPPGESGA A
This gene encodes the pro-alpha2 chain of type I collagen whose triple helix comprises two alpha1 chains and one alpha2 chain. Type I is a fibril-forming collagen found in most connective tissues and is abundant in bone, cornea, dermis and tendon. Mutations in this gene are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIB, recessive Ehlers-Danlos syndrome Classical type, idiopathic osteoporosis, and atypical Marfan syndrome. Symptoms associated with mutations in this gene, however, tend to be less severe than mutations in the gene for the alpha1 chain of type I collagen (COL1A1) reflecting the different role of alpha2 chains in matrix integrity. Three transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-2 type I collagen; COL1; Collagen alpha-2(I) chain; collagen COL1A2; Collagen I; osteogenesis imperfecta; procollagen, type I, alpha 2
Gene Aliases: AA960264; AI325291; Col1a-2; Col1a2; Cola-2; Cola2; oim
UniProt ID: (Mouse) Q01149, (Rat) P02466
Entrez Gene ID: (Mouse) 12843, (Rat) 84352
Molecular Function:
extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support