Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EEMVQNHMT YSLQDVGGDA NWQLVVEEGE MKVYRREVEE NGIVLDPLKA THAVKGVTGH EVCNYFWNVD VRNDWETTIE NFHVVETLAD NAIIIYQTHK RVWPASQRDV LYLSVIRKIP ALTENDPETW IVCNFSVDHD SAPLNNRCVR AKINVAMICQ TLVSPPEGNQ EISRDNILCK ITYVANVNPG GWAPASVLRA VAKREYPKFL KRFTSYVQEK TAGKPILF (398-624 aa encoded by BC000102)
CERT (GPBP) is a GC-rich promotor-specific transactivating transcription factor. This protein is suggested to play a role in the development of atherosclerosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha 3 type IV collagen binding protein; Ceramide transfer protein; ceramide transport protein; ceramide transporter; CERT; Collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; Goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; procollagen, type IV, alpha 3 (Goodpasture antigen) binding protein; StAR-related lipid transfer (START) domain containing 11; StAR-related lipid transfer protein 11; StARD11; START domain-containing protein 11
Gene Aliases: 2810404O15Rik; 9230101K08Rik; AU016711; CERT; CERT1; CERTL; COL4A3BP; GPBP; MRD34; STARD11
UniProt ID: (Human) Q9Y5P4, (Mouse) Q9EQG9
Entrez Gene ID: (Human) 10087, (Mouse) 68018, (Rat) 365652
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support