Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AYQLAVEAES EQKWKQLAEL AISKCQFGLA QECLHHAQDY GGLLLLATAS GNANMVNKLA EGAERDGKNN VAFMSYFLQG KVDACLELLI RTGRLPEAAF LARTYLPSQV SRVVKLWREN LSKVNQKAAE SLADPTEYEN LFPGLKEAFV VEEWVKETHA DLWPAKQYPL VTPNEERNVM EEGKDFQPSR STAQQELDGK PASPTPVIVA SHTANKEEKS LLELEVDLDN LELEDIDTTD INLDEDILDD; Positive Samples: SW480, MCF7, HeLa, HepG2, Jurkat, Mouse liver, Mouse kidney, Mouse stomach; Cellular Location: COPI-coated vesicle membrane, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle, Golgi apparatus membrane, Peripheral membrane protein, cytosol
The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non- clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network. Coatomer complex is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. In mammals, the coatomer can only be recruited by membranes associated to ADP-ribosylation factors (ARFs), which are small GTP-binding proteins; the complex also influences the Golgi structural integrity, as well as the processing, activity, and endocytic recycling of LDL receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta prime COP; Beta'-coat protein; Beta'-COP; betaprime-COP; coatomer binding complex, beta prime subunit; coatomer protein complex subunit beta 2 (beta prime); coatomer protein complex subunit beta prime; coatomer protein complex, subunit beta 2 (beta prime); Coatomer subunit beta'; p102
Gene Aliases: AI256832; beta'-COP; COPB2
UniProt ID: (Human) P35606, (Rat) O35142, (Mouse) O55029
Entrez Gene ID: (Human) 9276, (Rat) 60384, (Mouse) 50797
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support