Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ALTKQLEAV SDRVTVLYRS KAIRYTWPCP FPMADSSPWV HITLGDGSTF QTKLLIGADG HNSGVRQAVG IQNVSWNYDQ SAVVATLHLS EATENNVAWQ RFLPSGPIAL LPLSDTLSSL VWSTSHEHAA ELVSMDEEKF VDAVNSAFWS DADHTDFIDT AGAMLQYAVS LLKPTKVSAR QLPPSVARVD AKSRVLFPLG LGHAAEYVRP RVALIGDAAH RVHPLAGQGV NMGFGDISSL AHHLSTAAFN GKDLGSVSHL TGYETERQRH NTALLAATDL LKRLYSTSAS PLVLLRTWGL QATNAVSPLK EQIMAFASK (151-468 aa encoded by BC014181)
COQ6 (Coenzyme Q10 monooxygenase 6) is also named as CGI-10 and belongs to the ubiH/COQ6 family. It is an evolutionarily conserved flavin-dependent monooxygenase required for biosynthesis of coenzyme Q10, or ubiquinone. It has 3 isoforms with the molecular mass of 51 kDa, 43 kDa and 49 kDa. In inner ear, Coq6 is expressed in spiral ganglion, stria vascularis cells, and spiral ligament. Defects in COQ6 are the cause of coenzyme Q10 deficiency, primary, type 6 (COQ10D6).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coenzyme Q10 monooxygenase 6; coenzyme Q6 homolog, monooxygenase; coenzyme Q6 monooxygenase; ubiquinone biosynthesis monooxygenase COQ6; Ubiquinone biosynthesis monooxygenase COQ6, mitochondrial
Gene Aliases: 5930427M12Rik; AW742344; CGI-10; CGI10; COQ10D6; COQ6
UniProt ID: (Human) Q9Y2Z9, (Rat) Q68FU7, (Mouse) Q8R1S0
Entrez Gene ID: (Human) 51004, (Rat) 299195, (Mouse) 217707
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support