Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: EEWFEGKNAD PILISLKHGY IPGKNRDLKV VKKNILDSKP TANKKCDLIS IPKKTTDTAS VQNEAKLDEI LKEIKSIKDT ICNQDERISK LEQQMAKIA
Coronin 3 is associated with the aggression and metastasis of hepatocellular carcinoma. Coronins are a family of highly evolutionary conserved proteins reportedly involved in the regulation of actin cytoskeletal dynamics, although only coronin 3 has been shown to be related to cancer cell migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coronin, actin binding protein, 1C; coronin, actin-binding protein, 1C; Coronin-1C; Coronin-3; hCRNN4
Gene Aliases: CORO1C; CRN2; CRNN4; HCRNN4
UniProt ID: (Human) Q9ULV4
Entrez Gene ID: (Human) 23603
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support