Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAASPHTLSS RLLTGCVGGS VWYLERRTIQ DSPHKFLHLL RNVNKQWITF QHFSFLKRMY VTQLNRSHNQ QVRPKPEPVA SPFLEKTSSG QAKAEIYEMR PLSPPSLSLS RKPNEKELIE LEPDSVIEDS IDVGKETKEE KRWKEMKLQV YDLPGILAQL SKIKLTALVV STTAAGFALA PGPFDWPCFL LTSVGTGLAS CAANSINQFF EVPFDSNMNR TKNRPLVRGQ ISPLLAVSFA TCCAVPGVAI LTLGVNPLTG ALGLFNIFLY TCCYTPLKRI SIANTWVGAV VGAIPPVMGW TAATGSLDAG AFLLGGILYS WQFPHFNALS WGLREDYSRG GYCMMSVTHP GLCRRVALRH CLALLVLSAA APVLDITTWT FPIMALPINA YISYLGFRFY VDADRRSSRR LFFCSLWHLP LLLLLMLTCK RPSGGGDAGP PPS
Defects in COX10 are a cause of mitochondrial complex IV deficiency (MT-C4D); also known as cytochrome c oxidase deficiency. A disorder of the mitochondrial respiratory chain with heterogeneous clinical manifestations, ranging from isolated myopathy to severe multisystem disease affecting several tissues and organs. Features include hypertrophic cardiomyopathy, hepatomegaly and liver dysfunction, hypotonia, muscle weakness, excercise intolerance, developmental delay, delayed motor development and mental retardation. A subset of patients manifest Leigh syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COX10 heme A:farnesyltransferase cytochrome c oxidase assembly factor; COX10 homolog, cytochrome c oxidase assembly protein, heme A: farnesyltransferase; cytochrome c oxidase assembly homolog 10; cytochrome c oxidase assembly protein; cytochrome c oxidase subunit X; heme A: farnesyltransferase; Heme O synthase; Protoheme IX farnesyltransferase, mitochondrial
Gene Aliases: COX10
UniProt ID: (Human) Q12887
Entrez Gene ID: (Human) 1352
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support