Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: LTESGLSMVD WHLIKEMKPP TSQEEWEAEF QRYQQFPEFK ILNHDMTLTE FKFIWYMEYS H
The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX15 (cytochrome c oxidase assembly protein COX15 homolog) is a 410 amino acid multi-pass mitochondrial membrane protein belonging to the COX15/ctaA family. Existing as two alternatively spliced isoforms, COX15 is predominantly expressed in tissues containing high rates of oxidative phosphorylation including brain, heart and muscle. COX15 defects are the cause of a neurodegenerative disorder known as Leigh syndrome and cytochrome c oxidase deficiency (COX deficiency).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COX15 cytochrome c oxidase assembly homolog; COX15 homolog, cytochrome c oxidase assembly protein; cytochrome c oxidase assembly homolog 15; Cytochrome c oxidase assembly protein COX15 homolog; cytochrome c oxidase subunit 15; HAS; Heme A synthase COX15
Gene Aliases: CEMCOX2; COX15
UniProt ID: (Human) Q7KZN9
Entrez Gene ID: (Human) 1355
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support