Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Cytochrome c oxidase, the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIc, which has 77% amino acid sequence identity with mouse subunit VIc. This gene is up-regulated in prostate cancer cells. A pseudogene has been found on chromosomes 16p12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytochrome c oxidase polypeptide VIc; Cytochrome c oxidase polypeptide VIc-2; Cytochrome c oxidase subunit 6C; Cytochrome c oxidase subunit 6C-2; cytochrome c oxidase subunit VIc preprotein; cytochrome oxidase subunit VIc; EC 1.9.3.1
Gene Aliases: COVIc; COX-VIc; COX6C; Cox6c2
UniProt ID: (Human) P09669, (Mouse) Q9CPQ1, (Rat) P11951
Entrez Gene ID: (Human) 1345, (Mouse) 12864, (Rat) 54322
Molecular Function: oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support