Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: DAQFTQRKAE RATLRSHFRD KYRLPKNETD ESQIQMAGGD VELPRELAKM IEEDTEEEEE KASVLGQLAS LPGLNLGSLK DKAQATLGDL KQSAEKCHVM; Positive Samples: Mouse brain, Mouse eye, Rat brain; Cellular Location: Cell junction, Lipid-anchor, Membrane, synapse
Positively regulates a late step in synaptic vesicle exocytosis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complexin III; Complexin-3; CPX III; lectin, mannose-binding, 1 like
Gene Aliases: BC016632; Cplx3; CpxIII; ERGIC-53L; ERGL; Lamn1l; Lman1l; RGD1560104
UniProt ID: (Mouse) Q8R1B5
Entrez Gene ID: (Mouse) 235415, (Rat) 501005
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support