Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HVQRAEMLP KTSGTRATSL GRPEEEEDEL AHRCSSFMAP PVTDLGELRR RPGDMKTKME LLILETQAQV CQALAQVDGG ANFSVDRWER KEGGGGISCV LQDGCVFEKA GVSISVVHGN LSEEAAKQMR SRGKVLKTKD GKLPFCAMGV SSVIHPKNPH APTIHFNYRY FEVEEADGHK QWWFGGGCDL TPTYLNQEDA VHFHRTLKEA CDQHGPDLYP KFKKWCDDYF FIAHRGERRG IGGIFFDDLD SPSKEEVFRF VQSCARAVVP SYIPLVKKHC DDSFTPQEKL WQQLRRGRYV EFNLLYDRGT KFGLFTPGSR IESILMSLPL TARWEYMHSP SENSKEAEIL EVLRHPRDWV R (95-454 aa encoded by BC017210)
The protein encoded by this gene is the sixth enzyme of the heme biosynthetic pathway. The encoded enzyme is soluble and found in the intermembrane space of mitochondria. This enzyme catalyzes the stepwise oxidative decarboxylation of coproporphyrinogen III to protoporphyrinogen IX, a precursor of heme. Defects in this gene are a cause of hereditary coproporphyria (HCP).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: clone 560; coprogen oxidase; Coproporphyrinogen III oxidase, mitochondrial precursor (Coproporphyrinogenase) (Coprogen oxidase) (COX); coproporphyrinogen-III oxidase, mitochondrial; coproporphyrinogenase; COX; cpox {ECO:0000312|EMBL:AAI07665.1}; Oxygen-dependent coproporphyrinogen-III oxidase, mitochondrial
Gene Aliases: cac; CPO; CPOX; CPX; HCP; nct
UniProt ID: (Human) P36551, (Rat) Q3B7D0, (Mouse) Q8VD08
Entrez Gene ID: (Human) 1371, (Rat) 304024, (Mouse) 12892
Molecular Function: oxidase oxidoreductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support