Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SRELFESWC TDKRNGVIIA GYCVEGTLAK HIMSEPEEIT TMSGQKLPLK MSVDYISFSA HTDYQQTSEF IRALKPPHVI LVHGEQNEMA RLKAALIREY EDNDEVHIEV HNPRNTEAVT LNFRGEKLAK VMGFLADKKP EQGQRVSGIL VKRNFNYHIL SPCDLSNYTD LAMSTVKQTQ AIPYTGPFNL LCYQLQKLTG DVEELEIQEK PALKVFKNIT VIQEPGMVVL EWLANPSNDM YADTVTTVIL EVQSNPKIRK GAVQKVSKKL EMHVYSKRLE IMLQDIFGED CVSVKDDSIL SVTVDGKTAN LNLETRTVEC EEGSEDDESL REMVELAAQR LYEALTPVH (337-684 aa encoded by BC020211)
Most eukaryotic mRNA precursors (pre-mRNAs) undergo extensive maturational processing, including cleavage and polyadenylation at the 3-prime end. CPSF3 encodes the 73-kD subunit of the cleavage and polyadenylation specificity factor (CPSF) and is the pre-mRNA 3-prime-end-processing endonuclease (Mandel et al., 2006).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cleavage and polyadenylation specific factor 3, 73kDa; Cleavage and polyadenylation specificity factor 73 kDa subunit; Cleavage and polyadenylation specificity factor subunit 3; CPSF; CPSF 73 kDa subunit; mRNA 3'-end-processing endonuclease CPSF-73; YSH1
Gene Aliases: CPSF-73; CPSF3; CPSF73
UniProt ID: (Human) Q9UKF6
Entrez Gene ID: (Human) 51692
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support