Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Most eukaryotic mRNA precursors (pre-mRNAs) undergo extensive maturational processing, including cleavage and polyadenylation at the 3-prime end. CPSF3 encodes the 73-kD subunit of the cleavage and polyadenylation specificity factor (CPSF) and is the pre-mRNA 3-prime-end-processing endonuclease (Mandel et al., 2006).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cleavage and polyadenylation specific factor 3, 73kDa; cleavage and polyadenylation specificity factor 3; Cleavage and polyadenylation specificity factor 73 kDa subunit; Cleavage and polyadenylation specificity factor subunit 3; CPSF; CPSF 73 kDa subunit; mRNA 3'-end-processing endonuclease CPSF-73; YSH1
Gene Aliases: CPSF-73; CPSF3; CPSF73
UniProt ID: (Human) Q9UKF6, (Mouse) Q9QXK7
Entrez Gene ID: (Human) 51692, (Mouse) 54451, (Rat) 298916
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support