Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, X. laevis, X. tropicalis and Bovine
Inhibition of the nuclear export of poly(A)-containing mRNAs caused by the influenza A virus NS1 protein requires its effector domain. The NS1 effector domain functionally interacts with the cellular 30 kDa subunit of cleavage and polyadenylation specific factor 4, an essential component of the 3' end processing machinery of cellular pre-mRNAs. In influenza virus-infected cells, the NS1 protein is physically associated with cleavage and polyadenylation specific factor 4, 30kD subunit. Binding of the NS1 protein to the 30 kDa protein in vitro prevents CPSF binding to the RNA substrate and inhibits 3' end cleavage and polyadenylation of host pre-mRNAs. Thus the NS1 protein selectively inhibits the nuclear export of cellular, and not viral, mRNAs. Multiple alternatively spliced transcript variants that encode different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cleavage and polyadenylation specific factor 4, 30kD subunit; cleavage and polyadenylation specific factor 4, 30kDa; Cleavage and polyadenylation specificity factor 30 kDa subunit; Cleavage and polyadenylation specificity factor subunit 4; Clipper homolog; Clipper/CPSF 30K; CPSF 30 kDa subunit; musculus clipper/cleavage and polyadenylation specificity factor 30 kDa subunit; Neb-1; no; No arches homolog; no arches-like zinc finger protein; NS1 effector domain-binding protein 1
Gene Aliases: 30kDa; C79664; CPSF30; CPSF4; NAR; NEB-1; NEB1
UniProt ID: (Human) O95639, (Mouse) Q8BQZ5
Entrez Gene ID: (Human) 10898, (Mouse) 54188
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support