Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QIESSKGTNK EGGGSANTRG ASTPPTLGDL FAGGFPVLRP AGQRDVAGGK TGQGPGSRAP SPRLPNKTIS GPLIPPASPR LGNTS
Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 85%.
May be a regulator of cytoskeletal organization. May have a role in spermatogenesis. Target protein binds SH3 domains; may play a role in MAP kinase mediated signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Corticosteroids and regional expression protein 16 homolog; FLJ36931; WAS/WASL interacting protein family, member 3; WAS/WASL-interacting protein family member 3
Gene Aliases: CR16; WIPF3
UniProt ID: (Human) A6NGB9
Entrez Gene ID: (Human) 644150
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support