Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody can bind the close sequences genes.
Immunogen sequence: MLAFAARTV VKPLGFLKPF SLMKASSRFK AHQDALPRLP VPPLQQSLDH YLKALQPIVS EEEWAHTKQL VDEFQASGGV GERLQKGLER RARKTENWLS EWWLKTAYLQ YRQPVVIYSS PGVMLPKQDF VDLQGQLRFA AKLIEGVLDF KVMIDNETLP VEYLGGKPLC MNQYYQILSS CRVPGPKQDT VSNFSKTKKP PTHITVVHNY QFFELDVYHS DGTPLTADQI FVQLEKIWNS SLQTNKEPVG ILTSNHRNSW AKAYNTLIKD KVNRDSVRSI QK (1-281 aa encoded by BC000723)
Carnitine acetyltransferase (CRAT) is a key enzyme in the metabolic pathway in mitochondria, peroxisomes and endoplasmic reticulum. CRAT catalyzes the reversible transfer of acyl groups from an acyl-CoA thioester to carnitine and regulates the ratio of acylCoA/CoA in the subcellular compartments. Different subcellular localizations of the CRAT mRNAs are thought to result from alternative splicing of the CRAT gene suggested by the divergent sequences in the 5' region of peroxisomal and mitochondrial CRAT cDNAs and the location of an intron where the sequences diverge. The alternatively splicing of this gene results in three distinct isoforms, one of which contains an N-terminial mitochondrial transit peptide, and has been shown to be located in mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carnitine acetylase; Carnitine acetyltransferase; Carnitine O-acetyltransferase; CAT
Gene Aliases: AW107812; CARAT; CAT1; CRAT
UniProt ID: (Human) P43155, (Rat) Q704S8, (Mouse) P47934
Entrez Gene ID: (Human) 1384, (Rat) 311849, (Mouse) 12908
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support