Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
Sequence Similarities: 95% Mouse/Rat.
Tissue Specificity: Widely expressed. Highly expressed in brain.
Positive Control: HEK-293 cell lysate, A375 cell lysate, A549 cell lysate, HeLa cell lysate, SH-SY5Y cell lysate, NIH/3T3 cell lysate, MCF7 cell lysate, mouse brain tissue lysate, rat testis tissue lysate, human testis tissue, human brain tissue.
Subcellular Location: Cytoplasm, Nucleus, Membrane.
Predicted band size: 51 kDa.
CRBN, a member of the Lon protease protein family, plays a role in brain development. It is a component of the DCX (DDB1-CUL4-X-box) E3 protein ligase complex, a complex that mediates the ubiquitination and subsequent proteasomal degradation of target proteins and is required for limb outgrowth and expression of the fibroblast growth factor FGF8. CRBN is thought to regulate the assembly and neuronal surface expression of large-conductance calcium-activated potassium channels in brain regions involved in memory and learning via its interaction with KCNT1. It is widely expressed and highly expressed in brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781K0715; MGC27358; novel lethal; Protein cereblon; Protein PiL; protein x 0001
Gene Aliases: 2610203G15Rik; 2900045O07Rik; AD-006; AF229032; AW108261; CRBN; MRT2; MRT2A; piL
UniProt ID: (Human) Q96SW2, (Mouse) Q8C7D2, (Rat) Q56AP7
Entrez Gene ID: (Human) 51185, (Mouse) 58799, (Rat) 297498
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support