Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NQMLEAQEE HLEAWWLQLK SEYPDLFEWF CVKTLKVCCS PGTYGPDCLA CQGGSQRPCS GNGHCSGDGS RQGDGSCRCH MGYQGPLCTD CMDGYFSSLR NETHSICTAC DESCKTCSGL TNRDCGECEV GWVLDEG (91-226 aa encoded by BC050675)
CRELD2 may regulate transport of alpha4-beta2 neuronal acetylcholine receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CREL2; cysteine rich with EGF-like domains 2; Cysteine-rich with EGF-like domain protein 2; cysteine-rich with EGF-like domains 2; DKFZp667O055; MGC11256; Protein disulfide isomerase CRELD2
Gene Aliases: CRELD2; UNQ185/PRO211
UniProt ID: (Human) Q6UXH1
Entrez Gene ID: (Human) 79174
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support