Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRLPRRAALG LLPLLLLLPP APEAAKKPTP CHRCRGLVDK FNQGMVDTAK KNFGGGNTAW EEKTLSKYES SEIRLLEILE GLCESSDFEC NQMLEAQEEH LEAWWLQLKS EYPDLFEWFC VKTLKVCCSP GTYGPDCLAC QGGSQRPCSG NGHCSGDGSR QGDGSCRCHM GYQGPLCTDC MDGYFSSLRN ETHSICTACD ESCKTCSGLT NRDCGECEVG WVLDEGACVD VDECAAEPPP CSAAQFCKNA NGSYTCEDVD ECSLAEKTCV RKNENCYNTP GSYVCVCPDG FEETEDACVP PAEAEATEGE SPTQLPSRED L
CRELD2 may regulate transport of alpha4-beta2 neuronal acetylcholine receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CREL2; cysteine rich with EGF-like domains 2; Cysteine-rich with EGF-like domain protein 2; cysteine-rich with EGF-like domains 2; DKFZp667O055; MGC11256; Protein disulfide isomerase CRELD2
Gene Aliases: CRELD2; UNQ185/PRO211
UniProt ID: (Human) Q6UXH1
Entrez Gene ID: (Human) 79174
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support