Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSPNFKLQCH FILIFLTALR GESRYLELRE AADYDPFLLF SANLKRELAG EQPYRRALRC LDMLSLQGQF TFTADRPQLH CAAFFISEPE EFITIHYDQV SIDCQGGDFL KVFDGWILKG EKFPSSQDHP LPSAERYIDF CESGLSRRSI RSSQNVAMIF FRVHEPGNGF TLTIKTDPNL FPCNVISQTP NGKFTLVVPH QHRNCSFSII YPVVIKISDL TLGHVNGLQL KKSSAGCEGI GDFVELLGGT GLDPSKMTPL ADLCYPFHGP AQMKVGCDNT VVRMVSSGKH VNRVTFEYRQ LEPYELENPN GNSIGEFCLS GL
Response to stress in mammals requires an intact hypothalamic-pituitary-adrenal axis. The proximal part of the response is mediated by secretion of corticotropin-releasing hormone (CRH) by the paraventricular nucleus of the hypothalamus. CRH is a 41 amino acid peptide derived by enzymatic cleavage from a 191 amino acid preprohormone. CRH is produced not only in the hypothalamus but also in peripheral tissues, such as T lymphocytes; it is highly expressed in human placenta. Glucocorticoids stimulate placental CRH synthesis and secretion in primary cultures of human placenta. This stimulation is in contrast to the glucocorticoid suppression of CRH expression in hypothalamus. The gene which encodes CRH maps to human chromosome 8q13. Human plasma contains a CRH-binding protein, CRH-BP (also designated CRF-BP) which inactivates CRH and which may prevent inappropriate pituitary-adrenal stimulation in pregnancy. The gene which encodes CRF-BP maps to human chromosome 5q11.2-q13.3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: corticotropin releasing hormone-binding protein; Corticotropin-releasing factor-binding protein; Corticotropin-releasing hormone-binding protein; CRF-binding protein; CRF-BP; CRH-BP
Gene Aliases: CRF-BP; CRFBP; CRHBP
UniProt ID: (Human) P24387
Entrez Gene ID: (Human) 1393
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support