Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: WSKNWAKAA AFVTSPPLSP DPTTPDYLTS LLACGDLQVT GSGHCPYSTA QKAVGKDNFT LIPEGVNGIE ERMTVVWDKA VATGKMDENQ FVAVTSTNAA KIFNLYPRKG RIAVGSDADV VIWDPDKLKT ITAKSHKSAV EYNIFEGMEC HGSPLVVISQ GKIVFEDGNI NVNKGMGRFI PRKAFPEHLY QRVKIRNKVF GLQGVSRGMY DGPVYEVPAT PKYATPAPSA KSSPSKHQPP PIRNLHQS (291-537 aa encoded by BC000252)
Collapsin-response mediator proteins (CRMPs) are highly expressed in the developing brain where they play major roles in axonal outgrowth, neurite differentiation, and apoptosis. Their continued expression in areas of high synaptic remodeling such as the cerebellum, hippocampus, and the olfactory system suggests that these proteins may also be involved in adult brain plasticity. CRMP-1 was initially identified as a dihydro-pyrimidinase expressed exclusively in brain; later studies have shown that it is involved with neurotrophin (NT) 3-induced neurite formation and outgrowth. CRMP-1 localization switches from axonal to somatodendritic when neurons reach functional maturity, suggesting that it is involved in early neuronal differentiation as well as in later processes related to the survival or death of the newly generated neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collapsin response mediator protein 1; CRMP-1; dihydropyrimidinase-like 1; Dihydropyrimidinase-related protein 1; DRP-1; Inactive dihydropyrimidinase; ULIP-3; Unc-33-like phosphoprotein 3
Gene Aliases: CRMP-1; CRMP1; DPYSL1; DRP-1; DRP1; ULIP-3; ULIP3
UniProt ID: (Human) Q14194, (Rat) Q62950, (Mouse) P97427
Entrez Gene ID: (Human) 1400, (Rat) 25415, (Mouse) 12933
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support