Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EGHVLSRPE ELEAEAVFRA ITIASQTNCP LYVTKVMSKS AADLISQARK KGNVVFGEPI TASLGIDGTH YWSKNWAKAA AFVTSPPLSP DPTTPDYINS LLASGDLQLS GSAHCTFSTA QKAIGKDNFT AIPEGTNGVE ERMSVIWDKA VATGKMDENQ FVAVTSTNAA KIFNLYPRKG RISVGSDSDL VIWDPDAVKI VSAKNHQSAA EYNIFEGMEL RGAPLVVICQ GKIMLEDGNL HVTQGAGRFI PCSPFSDYVY KRIKARRKMA DLHAVPRGMY DGPVFDLTTT PKGGTPAGSA RGSPTRPNPP VRNLHQSGFS LSGTQVDEGV RSASKRIVAP PGGRSNITSL S (221-570 aa encoded by BC039006)
DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collapsin response mediator protein 4; collapsin response mediator protein 4 long; CRMP-4; dihydropyrimidinase-like 3; Dihydropyrimidinase-related protein 3; DRP-3; testicular secretory protein Li 7; TOAD-64/Ulip/CRMP; TOAD-64/Ulip/CRMP protein 4b; tuc-4; TUC-4b; ULIP-1; Unc-33-like phosphoprotein 1
Gene Aliases: CRMP-4; CRMP4; DPYSL3; DRP-3; DRP3; LCRMP; TUC-4b; TUC4; ULIP; ULIP-1; ULIP1
UniProt ID: (Human) Q14195, (Mouse) Q62188, (Rat) Q62952
Entrez Gene ID: (Human) 1809, (Mouse) 22240, (Rat) 25418
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support