Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ASHSGIPNII LTVTGESPPS LSKELTSSLA GVGDVSFDSD SQFPLDELKI DPLTLDGLHM LNDPDMVLAD PATEDTFRMD RL
Transcriptional coactivator for CREB1 which activates transcription through both consensus and variant cAMP response element (CRE) sites. Acts as a coactivator, in the SIK/TORC signaling pathway, being active when dephosphorylated and acts independently of CREB1 'Ser-133' phosphorylation. Enhances the interaction of CREB1 with TAF4. Regulates the expression of specific CREB-activated genes such as the steroidogenic gene, StAR. Potent coactivator of PGC1alpha and inducer of mitochondrial biogenesis in muscle cells. In the hippocampus, involved in late-phase long-term potentiation (L-LTP) maintenance at the Schaffer collateral-CA1 synapses. May be required for dendritic growth of developing cortical neurons. In concert with SIK1, regulates the light-induced entrainment of the circadian clock. In response to light stimulus, coactivates the CREB-mediated transcription of PER1 which plays an important role in the photic entrainment of the circadian clock. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CREB-regulated transcription coactivator 1; CRTC1; FLJ14027; MECT 1; Mucoepidermoid carcinoma translocated protein 1; TORC-1; transducer of regulated cAMP response element-binding protein (CREB) 1; Transducer of regulated cAMP response element-binding protein 1
Gene Aliases: CRTC1; KIAA0616; MECT1; TORC-1; TORC1; WAMTP1
UniProt ID: (Human) Q6UUV9
Entrez Gene ID: (Human) 23373
Molecular Function: transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support