Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta c; beta common cytokine receptor; CD antigen CD131; CD131; CD131 antigen; CDw131; colony stimulating factor 2 receptor, beta 1, low-affinity (granulocyte-macrophage); colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage); Colony-stimulating factor 2 receptor subunit beta-2; colony-stimulating factor-2 receptor, beta, low-affinity; common beta chain; Cytokine receptor common subunit beta; GM-CSF/IL-3/IL-5 receptor common beta subunit; GM-CSF/IL-3/IL-5 receptor common beta-chain; IL-3 RB; IL-3 Receptor beta; IL-3 receptor class 2 subunit beta; IL-3R class 2 subunit beta; IL3R beta; interleukin 3 common beta chain; interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3 receptor, beta (high affinity); Interleukin-3 receptor class 2 subunit beta; Interleukin-3 receptor class II beta chain
Gene Aliases: Ai2ca; AI848964; AIC2A; AIC2B; Bc; BetaIl3; Bil3; CD131; CDw131; CSF2RB; Csf2rb1; Csf2rb2; Csfgmrb; Il3r; IL3RB; Il3rb1; Il3rb2; IL5RB; SMDP5
UniProt ID: (Human) P32927, (Mouse) P26955, (Mouse) P26954
Entrez Gene ID: (Human) 1439, (Rat) 171081, (Mouse) 12983, (Mouse) 12984
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support