Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total CSGLCAT.
CSGlcA-T belongs to the chondroitin N-acetylgalactosaminyltransferase family. It transfers glucuronic acid (GlcUA) from UDP-GlcUA to N-acetylgalactosamine residues on the non-reducing end of the elongating chondroitin polymer. CSGlcA-T has no N-acetylgalactosaminyltransferase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chondroitin glucuronyltransferase; Chondroitin polymerizing factor 2; Chondroitin sulfate glucuronyltransferase; Chondroitin synthase 3; chondroitin synthase-3; ChPF-2; ChSy-3; CSGlcA-T; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase
Gene Aliases: chPF-2; CHPF2; ChSy-3; CHSY3; CSGLCA-T; CSGLCAT; KIAA1402; UNQ299/PRO339
UniProt ID: (Human) Q9P2E5
Entrez Gene ID: (Human) 54480
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support