Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COP9 signalosome complex subunit 1; G protein pathway suppressor 1; GPS-1; JAB1-containing signalosome subunit 1; MFH protein; MGC71287; Protein MFH; SGN1; signalosome subunit 1
Gene Aliases: COPS1; CSN1; GPS1; MFH; R75577; SGN1
UniProt ID: (Human) Q13098, (Rat) P97834
Entrez Gene ID: (Human) 2873, (Rat) 117039, (Mouse) 209318
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support