Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: SPKPARSVTT SNPSKFTAKF GESEKCPRCG KSVYAAEKVM GGGKPWHKTC FRCAICGKSL ESTNVTDKDG ELYCKVCYAK NFGPTGIGFG GLTQQVEKKE
This gene encodes a member of the CSRP family of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this protein is found in a group of proteins with critical functions in gene regulation, cell growth, and somatic differentiation. Mutations in this gene are thought to cause heritable forms of hypertrophic cardiomyopathy (HCM) and dilated cardiomyopathy (DCM) in humans. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac LIM domain protein; Cardiac LIM protein; CRP3; Cysteine and glycine-rich protein 3; cysteine and glycine-rich protein 3 (cardiac LIM protein); Cysteine-rich protein 3; LIM domain only 4; LIM domain protein, cardiac; MGC14488; MGC61993; Muscle LIM protein; muscle lim protein isoform
Gene Aliases: CLP; CMD1M; CMH12; CRP3; CSRP3; LMO4; MLP
UniProt ID: (Human) P50461
Entrez Gene ID: (Human) 8048
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support