Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CT45A3 (Cancer/Testis Antigen Family 45 Member A3) is a Protein Coding gene. An important paralog of this gene is CT45A1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 45-1; Cancer/testis antigen 45-3; Cancer/testis antigen 45-4; cancer/testis antigen 45-4/6; Cancer/testis antigen 45-5; Cancer/testis antigen 45-6; Cancer/testis antigen 45A1; Cancer/testis antigen 45A10; Cancer/testis antigen 45A3; Cancer/testis antigen 45A4; cancer/testis antigen 45A4/45A6; Cancer/testis antigen 45A5; Cancer/testis antigen 45A6; Cancer/testis antigen 45A7; cancer/testis antigen CT45-1; cancer/testis antigen CT45-3; cancer/testis antigen CT45-5; cancer/testis antigen CT45-6; cancer/testis antigen family 45 member A-like; Cancer/testis antigen family 45 member A1; Cancer/testis antigen family 45 member A10; Cancer/testis antigen family 45 member A3; Cancer/testis antigen family 45 member A4; cancer/testis antigen family 45 member A4/A6; Cancer/testis antigen family 45 member A5; Cancer/testis antigen family 45 member A6; Cancer/testis antigen family 45 member A7; cancer/testis antigen family 45, member A4
Gene Aliases: CT45; CT45-1; CT45-3; CT45-4; CT45-5; CT45-6; CT45.1; CT45.3; CT45.4; CT45.5; CT45.6; CT455; CT45A1; CT45A10; CT45A3; CT45A4; CT45A5; CT45A6; CT45A7
UniProt ID: (Human) Q5HYN5, (Human) P0DMU8, (Human) Q8NHU0
Entrez Gene ID: (Human) 541466, (Human) 102723631, (Human) 441519, (Human) 441521, (Human) 541465
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support