Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSRPQLRRW RLVSSPPSGV PGLALLALLA LRLAAGTDCP CPEPELCRPI RHHPDFEVFV FDVGQKTWKS YDWSQITTVA TFGKYDSELM CYAHSKGARV VLKGNL (1-105 aa encoded by B C024007)
Involved in the degradation of asparagine-linked glycoproteins. Hydrolyze of N-acetyl-beta-D-glucosamine (1-4)N-acetylglucosamine chitobiose core from the reducing end of the bond, it requires prior cleavage by glycosylasparaginase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chitobiase, di-N-acetyl-; CTBdi-N-acetylchitobiase; Di-N-acetylchitobiase
Gene Aliases: 2210401K11Rik; CTB; CTBS
UniProt ID: (Human) Q01459, (Rat) Q01460, (Mouse) Q8R242
Entrez Gene ID: (Human) 1486, (Rat) 81652, (Mouse) 74245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support