Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: GPCRCPDEPA PRCPAGVSLV LDGCGCCRVC AKQLGELCTE RDPCDPHKGL FCDFGSPANR KIGVCTAKDG APCIFGGTVY RSGESFQSSC KYQCTCLDGA VGCMPLCSMD VRLPSPDCPF PRRVKLPGKC CEEWVCDEPK DQTVVGPALA AYRLEDTFGP DPTMIRANCL VQTTEWSACS KTCGMGISTR VTNDNASCRL EKQSRLCMVR PCEADLEENI K
Connective Tissue Growth Factor (CTGF), also called CCN2, is a member of the CCN family. CTGF is a secreted protein produced by umbilical veins and vascular endothelial cells. CTGF possesses an Insulin-like Growth Factor (IGF)-binding domain, a thrombospondin type 1 domain, and a cysteine knot region. CTGF plays important roles in the proliferation and differentiation of chondrocytes, induces angiogenesis, and promotes cell adhesion in fibroblasts, endothelial, and epithelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-Nov; CCN family member 2; Cellular communication network factor 2; Connective tissue growth factor; H-CTGF; Hypertrophic chondrocyte-specific protein 24; IBP-8; IGF-binding protein 8; IGFBP-8; Insulin-like growth factor-binding protein 8; MGC102839
Gene Aliases: CCN2; CTGF; HCS24; IGFBP8; NOV2
UniProt ID: (Human) P29279
Entrez Gene ID: (Human) 1490
Molecular Function: growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support