Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody was raised against the N-terminal region of human cystinosin. Two isoforms of CTNS exist due to alternative splicing events and both of them can be detected through this antibody.
Immunogen sequence: VSLTVPPVV KLENGSSTNV SLTLRPPLNA TLVITFEITF RSKNITILEL PDEVVVPPGV TNSSFQVTSQ NVGQLTVYLH GNHSNQTGPR IRFLVIRSSA ISIINQVIGW IYFVAWSISF YPQVIMNWRR KSVIGLSFDF VALNLTGF (24-170 aa encoded by BC032850 )
This gene encodes a seven-transmembrane domain protein that functions to transport cystine out of lysosomes. Its activity is driven by the H+ electrochemical gradient of the lysosomal membrane. Mutations in this gene cause cystinosis, a lysosomal storage disorder. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cystinosin; cystinosis nephropathic; cystinosis, nephropathic
Gene Aliases: AI195360; AW049661; CTNS; CTNS-LSB; PQLC4
UniProt ID: (Human) O60931, (Mouse) P57757
Entrez Gene ID: (Human) 1497, (Mouse) 83429, (Rat) 287478
Molecular Function:
amino acid transporter
transporter
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support