Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 2
Product is supplied at 1 mL (100 µL/mL). Application Notes: Flow Cytometry - For intracellular staining activated plasmacytoid dendritic cells were fixed with 2% paraformaldehyde and permeabilized with 0.5% saponin. All steps were performed in presence of 0.5 M EDTA to avoid cell aggregation. Western Blot - Tissue was homogenized in loading sample buffer and subjected to SDS-PAGE. The band size is ~9 kDa. IHC-F - Tissue sections were fixed 10 minutes in acetone and pretreated with avidin/biotin blocking kit. As positive control chronically inflamed liver was used and as negative control normal liver and isotype matched antibody.
Positive control: Acitvated plasmacytoid dendritic cells; Negative control: Normal liver.
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 10 kDa interferon gamma-induced protein; C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 10; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; CXC; CXC motif chemokine; CXCL; gamma IP10; Gamma-IP10; H-IP-10; interferon-inducible cytokine IP-10; M-IP-10; protein 10 from interferon (gamma)-induced cell line; small inducible cytokine subfamily B (Cys-X-Cys), member 10; Small-inducible cytokine B10
Gene Aliases: C7; crg-2; CXCL10; gIP-10; IFI10; INP10; IP-10; mob-1; SCYB10
UniProt ID: (Human) P02778
Entrez Gene ID: (Human) 3627
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support