Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse CXCL17 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
CXCL17 is a chemokine that acts as chemoattractant for monocytes, macrophages and dendritic cells. In particular, CXCL17 is a mucosal chemokine that attracts dendritic cells and blood monocytes to the lungs. CXCL17 plays a role in angiogenesis and possibly in the development of tumors. Further, CXCL17 acts as an anti-inflammatory in the stomach and may play a role in the innate defense against infections. CXCL17 also activates the C-X-C chemokine receptor GPR35 to induce a rapid and transient rise in the level of intracellular calcium ions. 4-Cys CXCL17 seems to exhibit much higher chemoattractant potency on monocytes and macrophages than 6-Cys CXCL17. Diseases associated with CXCL17 dysfunction include cholera.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6-Cys CXCL17; C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 17; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 17; CXC; CXC motif chemokine; CXCL; Dendritic cell and monocyte chemokine-like protein; DMC; VEGF co-regulated chemokine 1; VEGF coregulated chemokine 1
Gene Aliases: CXCL17; Dcip1; DMC; UNQ473; UNQ473/PRO842; VCC-1; VCC1
UniProt ID: (Human) Q6UXB2
Entrez Gene ID: (Human) 284340
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support