Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total GRO gamma.
CXCL3 may play a role in inflammation and exert its effects on endothelial cells in an autocrine fashion. All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 3; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 3; CXC; CXC motif chemokine; CXCL; cxcl3; cytokine-induced neutrophil chemoattractant 2; cytokine-induced neutrophil chemoattractant-2; Dendritic cell inflammatory protein 1; gro gamma; GRO-gamma; GRO-gamma(1-73); GRO3 oncogene; Growth-regulated protein gamma; macrophage inflammatory protein 2-alpha/beta; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma; MIP2-alpha/beta; MIP2-beta
Gene Aliases: Cinc-2; CINC-2b; Cinc2; CXCL3; Dcip1; Gm1960; GRO3; GROG; MIP-2b; MIP2B; SCYB3
UniProt ID: (Human) P19876, (Mouse) Q6W5C0
Entrez Gene ID: (Human) 2921, (Mouse) 330122, (Rat) 171551
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support