Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: TPVVRKGRCS CISTNQGTIH LQSLKDLKQF APSPSCEKIE IIATLKNGVQ TCLNPDSADV KELIKKWEKQ VSQKKKQKNG KKHQKKKVLK VRKSQRSRQK KTT
MIG, also known as CXCL9, is a T-cell chemoattractant inducible by gamma interferon that is a member of the CXC chemokine family of cytokines.This gene is mapped to 4q21. It is noted that, although the best-described activities of the chemokines are as chemotactic factors, chemokines also have an effect on T-cell activation, angiogenesis, and HIV infection. While most CXC chemokines are chemotactic for neutrophils, MIG and INP10 are unusual and similar in being CXC chemokines that are chemotactic for lymphocytes and inactive in neutrophils.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 9; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CXC; CXC motif chemokine; CXCL; Gamma-interferon-induced monokine; HuMIG; monokine induced by gamma interferon; Monokine induced by interferon-gamma; Small-inducible cytokine B9
Gene Aliases: CMK; crg-10; CXCL9; Humig; MIG; SCYB9
UniProt ID: (Human) Q07325
Entrez Gene ID: (Human) 4283
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support