Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGAQLSTLG HMVLFPVWFL YSLLMKLFQR STPAITLESP DIKYPLRLID REIISHDTRR FRFALPSPQH ILGLPVGQHI YLSARIDGNL VVRPYTPISS DDDKGFVDLV IKVYFKDTHP KFPAGGKMSQ YLESMQIGDT IEFRGPSGLL VYQGKGKFAI RPDKKSNPII RTVKSVGMIA GGTGITPMLQ VIRAIMKDPD DHTVCHLLFA NQTEKDILLR PELEELRNKH SARFKLWYTL DRAPEAWDYG QGFVNEEMIR DHLPPPEEEP LVLMCGPPPM IQYACLPNLD HVGHPTERCF VF (1-301 aa encoded by BC004821)
CYB5R3 (NADH-cytochrome b5, reductase 3, diaphorase 1) is a 301 amino acid protein encoded by the human gene CYB5R3. CYB5R3 belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occuring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane, while isoform 2 is the soluble form found in erythrocytes. CYB5R3 is involved in the desaturation and elongation of fatty acids, cholesterol biosynthesis,drug metabolism and, in erythrocytes, methemoglobin reduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B5R; Diaphorase-1; Diaphorase1; mutant NADH-cytochrome b5 reductase; NADH-cytochrome b5 reductase 3; NADH-cytochrome b5 reductase 3 membrane-bound form; NADH-cytochrome b5 reductase 3 soluble form
Gene Aliases: B5R; CYB5R3; DIA1
UniProt ID: (Human) P00387
Entrez Gene ID: (Human) 1727
Molecular Function:
reductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support