Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAALGCARLR WALRGAGRGL CPHGARAKAA IPAALPSDKA TGAPGAGPGV RRRQRSLEEI PRLGQLRFFF QLFVQGYALQ LHQLQVLYKA KYGPMWMSYL GPQMHVNLAS APLLEQVMRQ EGKYPVRNDM ELWKEHRDQH DLTYGPFTTE GHHWYQLRQA LNQRLLKPAE AALYTDAFNE VIDDFMTRLD QLRAESASGN QVSDMAQLFY YFALEAICYI LFEKRIGCLQ RSIPEDTVTF VRSIGLMFQN SLYATFLPKW TRPVLPFWKR YLDGWNAIFS FGKKLIDEKL EDMEAQLQAA GPDGIQVSGY LHFLLASGQL SPREAMGSLP ELLMAGVDTT SNTLTWALYH LSKDPEIQEA LHEEVVGVVP AGQVPQHKDF AHMPLLKAVL KETLRLYPVV PTNSRIIEKE IEVDGFLFPK NTQFVFCHYV VSRDPTAFSE PESFQPHRWL RNSQPATPRI QHPFGSVPFG YGVRACLGRR IAELEMQLLL ARLIQKYKVV LAPETGELKS VARIVLVPNK KVGLQFLQRQ C
CYP27A1 participates in the degradation of cholesterol to bile acids in both the classic and acidic pathways. [1] It is the initiating enzyme in the acidic pathway to bile acids, yielding oxysterols by introducing a hydroxyl group to the carbon at the 27 position in cholesterol. In the acidic pathway, it produces 27-hydroxycholesterol from cholesterol whereas in the classic or neutral pathway, it produces 3beta-hydroxy-5-cholestenoic acid. It is also involved in the metabolism of vitamin D3. While CYP27A1 is present in many different tissues, its function in these tissues is largely uncharacterized. In macrophages, 27-hydroxycholesterol generated by this enzyme may be helpful against the production of inflammatory factors associated with cardiovascular disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 26-hydroxylase; 5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 27-hydroxylase; 5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 26-hydroxylase; 5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 27-hydroxylase; cholestanetriol 26-monooxygenase; cyt; Cytochrome P-450C27/25; Cytochrome P450 27; cytochrome P450, family 27, subfamily A, polypeptide 1; cytochrome P450, subfamily XXVIIA (steroid 27-hydroxylase, cerebrotendinous xanthomatosis), polypeptide 1; Sterol 26-hydroxylase, mitochondrial; Sterol 27-hydroxylase; Vitamin D(3) 25-hydroxylase
Gene Aliases: CP27; CTX; CYP27; CYP27A1
UniProt ID: (Human) Q02318
Entrez Gene ID: (Human) 1593
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support