Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
CYP27B1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein encoded by this gene localizes to the inner mitochondrial membrane where it hydroxylates 25-hydroxyvitamin D3 at the 1alpha position. This reaction synthesizes 1alpha,25-dihydroxyvitamin D3, the active form of vitamin D3, which binds to the vitamin D receptor and regulates calcium metabolism. Thus this enzyme regulates the level of biologically active vitamin D and plays an important role in calcium homeostasis. Mutations in this gene can result in vitamin D-dependent rickets type I.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1alpha(OH)ase; 25 hydroxyvitamin D3-1-alpha hydroxylase; 25(OH)D 1alpha-hydroxylase; 25-hydroxyvitamin D(3) 1-alpha-hydroxylase; 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial; 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial precursor (25-OHD-1 alpha-hydroxylase) (25-hydroxyvitamin D3 1-alpha-hydroxylase) (VD3 1A hydroxylase) (P450C1 alpha) (P450VD1-alpha); 25-hydroxyvitamin D3 1alpha-hydroxylase; 25-OHD-1 alpha-hydroxylase; Calcidiol 1-monooxygenase; cy; Cytochrome p450 27B1; cytochrome P450 40 (25-hydroxyvitamin D3 1 alpha-hydroxylase); cytochrome P450 subfamily XXVIIB (25-hydroxyvitamin D-1-alpha-hydroxylase) polypeptide 1; Cytochrome P450 subfamily XXVIIB polypeptide 1; cytochrome P450, 27b1; cytochrome P450, 40 (25-hydroxyvitamin D3 1 alpha-hydroxylase); cytochrome P450, family 27, subfamily B, polypeptide 1; cytochrome P450, subfamily 27b, polypeptide 1; Cytochrome P450C1 alpha; Cytochrome P450VD1-alpha; P450C1 alpha; P450VD1-alpha; P450VD1alpha; VD3 1A hydroxylase
Gene Aliases: CP2B; CYP1; CYP1ALPHA; CYP27B; CYP27B1; Cyp40; P450c1; PDDR; VDD1; VDDR; VDDRI; VDR
UniProt ID: (Human) O15528, (Mouse) O35084, (Rat) O35132
Entrez Gene ID: (Human) 1594, (Mouse) 13115, (Rat) 114700
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support