Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MALIPDLAME TWLLLAVSLV LLYLYGTHSH GLFKKLGIPG PTPLPFLGNI LSYHKGFCMF DMECHKKYGK VWGFYDGQQP VLAITDPDMI KTVLVKECYS VFTNRRPFGP VGFMKSAISI AEDEEWKRLR SLLSPTFTSG KLKEMVPIIA QYGDVLVRNL RREAETGKPV TLKDVFGAYS MDVITSTSFG VNIDSLNNPQ DPFVENTKKL LRFDFLDPFF LSITVFPFLI PILEVLNICV FPREVTNFLR KSVKRMKESR LEDTQKHRVD FLQLMIDSQN SKETESHKAL SDLELVAQSI IFIFAGYETT SSVLSFIMYE LATHPDVQQK LQEEIDAVLP NKAPPTYDTV LQMEYLDMVV NETLRLFPIA MRLERVCKKD VEINGMFIPK GVVVMIPSYA LHRDPKYWTE PEKFLPERFS KKNKDNIDPY IYTPFGSGPR NCIGMRFALM NMKLALIRVL QNFSFKPCKE TQIPLKLSLG GLLQPEKPVV LKVESRDGTV SGA
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. This enzyme is involved in the metabolism of approximately half the drugs in use today, including acetaminophen, codeine, cyclosporin A, diazepam and erythromycin. The enzyme also metabolizes some steroids and carcinogens. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Previously another CYP3A gene, CYP3A3, was thought to exist; however, it is now thought that this sequence represents a transcript variant of CYP3A4. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,4-cineole 2-exo-monooxygenase; 1,8-cineole 2-exo-monooxygenase; albendazole monooxygenase; Albendazole monooxygenase (sulfoxide-forming); Albendazole sulfoxidase; Cholesterol 25-hydroxylase; CYP; CYPIIIA3; CYPIIIA4; Cytochrome P450 3A3; Cytochrome P450 3A4; Cytochrome P450 HLp; Cytochrome P450 NF-25; cytochrome P450, family 3, subfamily A, polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; Cytochrome P450-PCN1; glucocorticoid-inducible P450; MGC126680; Nifedipine oxidase; P450; P450-III, steroid inducible; P450-PCN1; Quinine 3-monooxygenase; taurochenodeoxycholate 6-alpha-hydroxylase
Gene Aliases: CP33; CP34; CYP3A; CYP3A3; CYP3A4; CYPIIIA3; CYPIIIA4; HLP; NF-25; P450C3; P450PCN1
UniProt ID: (Human) P08684
Entrez Gene ID: (Human) 1576
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support