Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ILELSALVE KRSQHILQHM DFLYYLSHDG RRFHRACRLV HDFTDAVIRE RRRTLPTQGI DDFFKDKAKS KTLDFIDVLL LSKDEDGKAL SDEDIRAEAD TFMFGGHDTT ASGLSWVLYN LARHPEYQER CRQEVQELLK DRDPKEIEWD DLAQLPFLTM CVKESLRLHP PAPFISRCCT QDIVLPDGRV IPKGITCLID IIGVHHNPTV WPDPEVYDPF RFDPENSKGR SPLAFIPFSA GPRNCIGQAF AMAEMKVVLA LMLLHFRFLP DHTEPRRKLE LIMRAEGGLW LRVEPLNVSL Q (225-524 aa encoded by BC035350)
CYP4F12 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein likely localizes to the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYPIVF12; Cytochrome P450 4F12; cytochrome P450, family 4, subfamily F, polypeptide 12; cytochrome P450, subfamily IVF, polypeptide 12
Gene Aliases: CYP4F12; CYPIVF12; F22329_1; UNQ568/PRO1129
UniProt ID: (Human) Q9HCS2
Entrez Gene ID: (Human) 66002
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support