Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAAAGMLLL GLLQAGGSVL GQAMEKVTGG NLLSMLLIAC AFTLSLVYLI RLAAGHLVQL PAGVKSPPYI FSPIPFLGHA IAFGKSPIEF LENAYEKYGP VFSFTMVGKT FTYLLGSDAA ALLFNSKNED LNAEDVYSRL TTPVFGKGVA YDVPNPVFLE QKKMLKSGLN IAHFKQHVSI IEKETKEYFE SWGESGEKNV FEALSELIIL TASHCLHGKE IRSQLNEKVA QLYADLDGGF SHAAWLLPGW LPLPSFRRRD RAHREIKDIF YKAIQKRRQS QEKIDDILQT LLDATYKDGR PLTDDEVAGM LIGLLLAGQH TSSTTSAWMG FFLARDKTLQ KKCYLEQKTV CGENLPPLTY DQLKDLNLLD RCIKETLRLR PPVMIMMRMA RTPQTVAGYT IPPGHQVCVS PTVNQRLKDS WVERLDFNPD RYLQDNPASG EKFAYVPFGA GRHRCIGENF AYVQIKTIWS TMLRLYEFDL IDGYFPTVNY TTMIHTPENP VIRYKRRSK
P450 enzymes constitute a family of monooxygenase enzymes that are involved in the metabolism of a wide array of endogenous and xenobiotic compounds. Several P450 enzymes have been classified by sequence similarities as members of the CYP1A and CYP2A subfamilies. CYP51A1 (cytochrome P450, family 51, subfamily A, polypeptide 1), also known as LDM (lanosterol 14-alpha demethylase) or cytochrome P450-14DM, is a 503 amino acid protein localized to endoplasmic reticulum membrane. CYP51A1 is an important enzyme for zymosterol and steroid biosynthesis. CYP51A1 catalyzes C14-demethylation of lanosterol, transforming lanosterol into 4,4'-dimethyl cholesta-8,14,24-triene-3-beta-ol. CYP51A1 is ubiquitously expressed, with highest levels found in liver, ovary, testis, lung, kidney and prostate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYP51A1; CYPLI; Cytochrome P450 51A1; cytochrome P450, 51 (lanosterol 14-alpha-demethylase); cytochrome P450, family 51, subfamily A, polypeptide 1; Cytochrome P450-14DM; Cytochrome P45014DM; Cytochrome P450LI; Lanosterol 14-alpha demethylase; lanosterol 14-alpha-demethylase; LDM; P450 14DM; Sterol 14-alpha demethylase
Gene Aliases: CP51; CYP51; CYP51A1; CYPL1; LDM; P450-14DM; P450L1
UniProt ID: (Human) Q16850
Entrez Gene ID: (Human) 1595
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support