Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
1 µg/mL of MA5-45395 was sufficient for detection of Cav3.2 in 10 µg of HEK cell lysate expressing Cav3.2 by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 260kDa. No cross-reactivity against Cav1.3.
This gene encodes a T-type member of the alpha-1 subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. The alpha-1 subunit has 24 transmembrane segments and forms the pore through which ions pass into the cell. There are multiple isoforms of each of the proteins in the complex, either encoded by different genes or the result of alternative splicing of transcripts. Alternate transcriptional splice variants, encoding different isoforms, have been characterized for the gene described here. Studies suggest certain mutations in this gene lead to childhood absence epilepsy (CAE).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A1H-b; calcium channel alpha13.2 subunit; calcium channel, voltage-dependent, T type, alpha 1H subunit; calcium channel, voltage-dependent, T type, alpha 1Hb subunit; FLJ90484; Low-voltage-activated calcium channel alpha1 3.2 subunit; low-voltage-activated calcium channel alpha13.2 subunit; T-type Cav3.2; voltage dependent t-type calcium channel alpha-1H subunit; Voltage-dependent T-type calcium channel subunit alpha-1H; voltage-gated calcium channel alpha subunit Cav3.2; voltage-gated calcium channel alpha subunit CavT.2; Voltage-gated calcium channel subunit alpha Cav3.2
Gene Aliases: alpha13.2; CACNA1H; CACNA1HB; Cav3.2; ECA6; EIG6; MNCb-1209
UniProt ID: (Human) B5ME00, (Rat) Q9EQ60
Entrez Gene ID: (Human) 8912, (Rat) 114862, (Mouse) 58226
Molecular Function: voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support