Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TKKEFQATV EEGAVGVIVN LTVEDKDDPA TGAWRAAYTI INGNPGQSFE IHTNPQTNEG MLSVVKPLDY EISAFHTLLI KVENEDPLVP DVSYGPSSTA TVHITVLDVN EGPVFYPDPM MVTRQEDLSV GSVLLTVNAT DPDSLQHQTI RYSVYKDPAG WLNINPINGT VDTTAVLDRE STFVDNSVYT ALFLAIDSGN PPATGTGTLL ITLEDVNDNA PFIYPTVAEV CDDAKNLSVV ILGASDKDLH PNTDPFKFEI HKQAVPDKVW KISKINNTHA LVSLLQNLNK ANYNLPIMVT DSGKPPMTNI TDLRVQVCSC RNSKVDCNAA GALRFSLPSV LLLSLFSLAC L (364-713 aa encoded by BC030653)
The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3' trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAD13; cadherin 13, H-cadherin (heart); Cadherin-13; CD39-like 3; CD39L3; ecto-apyrase 3; ecto-ATPase 3; ecto-ATPDase 3; H-cadherin; H-cadherin (heart); Heart cadherin; heart-cadherin; NTPDase-3; P105; T cad; T Cadherin; T-cad; T-cadherin; Truncated cadherin; truncated-cadherin
Gene Aliases: 4932416G01Rik; CDH13; CDHH; Cdht; P105; T-cadherin; Tcad
UniProt ID: (Human) P55290, (Mouse) Q9WTR5
Entrez Gene ID: (Human) 1012, (Mouse) 12554
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support