Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Calcyon is a single transmembrane protein that interacts with D1 dopamine receptors. Dopamine is a neurotransmitter that regulates synaptic transmission involved in learning and memory. D1 receptors, the most abundant dopamine receptor in the central nervous system, appear to modulate the activity of D2 dopamine receptors, mediate various behavioural responses, and regulate neuron growth and differentiation. Calcyon is present in neuronal cell bodies and processes of the cortex and hippocampus, and it is especially abundant in pyramidal neurons. Interaction of Calcyon with D1 receptors results in a release of intracellular calcium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcyon D1 dopamine receptor-interacting protein (CALCYON); calcyon neuron-specific vesicular protein; calcyon protein; calcyon; D1 dopamine receptor-interacting protein; D1 dopamine receptor-interacting protein; dopamine receptor D1 interacting protein; Neuron-specific vesicular protein calcyon
Gene Aliases: 0710001P07Rik; 1110004A22Rik; Calcyon; CALY; DRD1IP; NSG3
UniProt ID: (Human) Q9NYX4, (Rat) P58821, (Mouse) Q9DCA7
Entrez Gene ID: (Human) 50632, (Rat) 192349, (Mouse) 68566
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support