Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 14
Keep as concentrated solution.
Predicted reactivity: Mouse (93%), Rat (93%), Zebrafish (82%), Xenopus laevis (89%), Dog (94%), Pig (94%), Chicken (91%), Bovine (94%).
Positive Control: mouse liver, PC-12, Rat2, 293T, A431, HeLa, HepG2.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Calnexin, also referred to as IP90, p88 and p90, is an approximately 90 kDa integral membrane protein of the endoplasmic reticulum (ER). Many resident ER proteins act as molecular chaperones and participate in the proper folding of polypeptides and their assembly into multisubunit proteins. Studies indicate that calnexin associates with the major histocompatibility complex (MHC) class I heavy chains, partial complexes of the T cell receptor and B cell membrane immunoglobulin, but not with completed receptor complexes. It has been shown that calnexin is a chaperone that retains incompletely or improperly folded proteins in the ER. The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble ER resident proteins such as GRP78 and GRP94 and protein disulfide isomerase. Integral membrane ER resident proteins, like calnexin, often lack this KDEL sequence but contain positively charged cytosolic residues that ensure ER retention. Calnexin contains a large ER luminal domain (461 amino acids), a transmembrane segment (22 amino acids), and a cytoplasmic tail (89 amino acids).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calnexin; FLJ26570; IP90; Major histocompatibility complex class I antigen-binding protein p88; p88; p90
Gene Aliases: 1110069N15Rik; AI988026; CANX; CNX; D11Ertd153e; IP90; P90
UniProt ID: (Human) P27824, (Rat) P35565, (Mouse) P35564
Entrez Gene ID: (Human) 821, (Rat) 29144, (Mouse) 12330
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support