Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Calpain proteinases are defined currently as papain-like neutral proteases that are calcium activated. Calpains 1 and 2 are composed of a large subunit, which is proteolytically active, and a small subunit (also called calpain-4), or enhancer protein, that is not proteolytically active. The other calpain proteins identified to date (calpain 5, 6, 8, 9, 10, 11, 12, 13, 14, 15) are not known to require a small subunit. Domains in the large subunit include the amino terminal domain-I, the proteinase domain-II, domain-III, and the EF-hand domain-IV (domain-T in calpains 5 & 6, domain-N for calpain-7). Calpain-7 domain-N does not have the standard calpain EF hand domain for binding calcium, making it unclear whether it is calcium regulated. In place of the EF hand calcium-binding domain IV, calpain-7 has a sequence most closely resembling domain-III, with unknown function. Calpain-7 is widely distributed, found in most tissues analyzed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calpain like protease; Calpain-7; homolog of Aspergillus Nidulans PALB; PalB homolog; PalBH
Gene Aliases: CALPAIN7; CAPN7; PALBH
UniProt ID: (Human) Q9Y6W3
Entrez Gene ID: (Human) 23473
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support