Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
The S100A8/A9 heterocomplex, also called MRP-8/MRP-14 complex or calprotectin, comprises 60% of the cytoplasmic protein fraction of circulating polymorphonuclear granulocytes and is also found in monocytes, macrophages and ileal tissue eosinophils. Peripheral blood monocytes carry the antigen extra- and intracellularly, neutrophils only intracellularly. The S100A8/A9 complex has antibacterial, antifungal, immunomodulating and antiproliferative effects. Besides this it is a potent chemotactic factor for neutrophils. Plasma concentrations are elevated in diseases associated with increased neutrophil activity, like inflammatory bowel disease. Granulocytes terminate their existence after transmigration through the intestinal wall. Therefore calprotectin is also detectable in feces. Elevated levels of calprotectin have been observed in body fluids such as plasma, saliva, gingival crevicular fluid, stools, and synovial fluid during infection and inflammatory conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AI323541; B8Ag; BEE11; BEE22; calcium binding protein; S100 protein; calgranulin A; calgranulin B; Calgranulin-A; Calgranulin-B; calprotectin; Calprotectin L1H subunit; Calprotectin L1L subunit; CFAG; Chemotactic cytokine CP-10; chemotactic S100 protein; Cystic fibrosis antigen; intracellular calcium-binding protein (MRP14); Leukocyte L1 complex heavy chain; Leukocyte L1 complex light chain; macrophage migration inhibitory factor-related protein-14; macrophage migration inhibitory factor-related protein-8; Migration inhibitory factor-related protein 14; Migration inhibitory factor-related protein 8; MRP-14; MRP-8; Myeloid-related protein 14; Neutrophil cytosolic 23 kDa protein; Neutrophil cytosolic 7 kDa protein; p14; p23; P7; p8; Pro-inflammatory S100 cytokine; Protein S100-A8; Protein S100-A9; S100 calcium binding protein A8; S100 calcium binding protein A8 (calgranulin A); S100 calcium binding protein A9 (calgranulin B); S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100A8/S100A9; Urinary stone protein band A
Gene Aliases: 60B8AG; AI323541; AW546964; B8Ag; BEE22; CAGA; CAGB; CFAG; CGLA; CGLB; CP-10; GAGB; L1AG; LIAG; MA387; MAC387; MIF; MRP-14; MRP-8; MRP14; MRP8; NIF; P14; P8; S100A8; S100A9
UniProt ID: (Bovine) P28782, (Human) P05109, (Bovine) P28783, (Rabbit) P50117, (Human) P06702, (Rat) P50115, (Mouse) P27005, (Mouse) P31725, (Rat) P50116
Entrez Gene ID: (Guinea pig) 100735514, (Horse) 100061766, (Dog) 490461, (Bovine) 616818, (Pig) 100127488, (Rabbit) 100008985, (Goat) 102168876, (Human) 6279, (Dog) 490463, (Bovine) 532569, (Pig) 100127489, (Horse) 100061730, (Rabbit) 100008704, (Goat) 102182273, (Human) 6280, (Rat) 116547, (Mouse) 20201, (Mouse) 20202, (Guinea pig) 100735238, (Rat) 94195
Molecular Function:
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support