Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Positive Control: Rat skeletal muscle tissue, mouse skeletal muscle tissue, human fetal skeletal muscle tissue, C2C12.
The sarcoplasmic reticulum (SR) is, in part, responsible for maintaining the level of intracellular calcium in cardiac and skeletal muscle by storing and releasing calcium. Several intralumenal SR calcium binding proteins have been identified, the most prominent of these is calsequestrin. Calsequstrin is a calcium binding protein known to sequester calcium accumulated in the sarcoplasmic reticulum of muscle cells during relaxation and is found discretely localized to the junctional and corbular (terminal cisternae) SR. Calsequestrin functions to localize calcium near the junctional face of the terminal cisternae from which calcium can be released into the cytosol via the ryanodine receptor. This protein is highly acidic and has a large capacity and moderate to low affinity for calcium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calmitin; Calmitine; calsequestrin 1 (fast-twitch, skeletal muscle); Calsequestrin, skeletal muscle isoform; Calsequestrin-1
Gene Aliases: CASQ; CASQ1; PDIB1; VMCQA
UniProt ID: (Human) P31415
Entrez Gene ID: (Human) 844
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support